missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-Naphthol-8-sulfonate 96.0+%, TCI America™

Catalog No. N003425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N003425G 25 g
N0034500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N003425G Supplier TCI America Supplier No. N003425G

Chemical Identifiers

CAS 20215-36-5
Molecular Formula C10H7NaO4S
Molecular Weight (g/mol) 246.212
MDL Number MFCD00021524
InChI Key PEYXCGCCIWJCTM-UHFFFAOYSA-M
Synonym 8-Hydroxy-1-naphthalenesulfonic Acid Sodium Salt, Sodium 8-Hydroxy-1-naphthalenesulfonate, 1-Naphthol-8-sulfonic Acid Sodium Salt
PubChem CID 23672328
IUPAC Name sodium;8-hydroxynaphthalene-1-sulfonate
SMILES C1=CC2=C(C(=C1)O)C(=CC=C2)S(=O)(=O)[O-].[Na+]

Specifications

Quantity 25 g
Formula Weight 246.21
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Naphthol-8-sulfonate
EINECSNumber (4)-0477
TSCA No