accessibility menu, dialog, popup

UserName

CAS RN 20215-36-5

IUPAC Name: sodium 8-hydroxynaphthalene-1-sulfonate
Synonyms: sodium 8-hydroxynaphthalene-1-sulfonate
Molecular Weight (g/mol): 246.21
Molecular Formula: C10H7NaO4S
InChi Key: PEYXCGCCIWJCTM-UHFFFAOYSA-M
SMILES: [Na+].OC1=C2C(C=CC=C2S([O-])(=O)=O)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results