missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-Naphthol-4-sulfonate 85.0+%, TCI America™

Supplier:  TCI America N0033100G

 View more versions of this product

Catalog No. N0033100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium 1-Naphthol-4-sulfonate
C10H7NaO4S
100 g
IAAKNVCARVEIFS-UHFFFAOYSA-M
sodium;4-hydroxynaphthalene-1-sulfonate
4626046
≥85.0% (HPLC,T)
6099-57-6
MFCD00021518
1-naphthol-4-sulfonic acid sodium salt, sodium naphthol sulfonate, sodium 1-naphthol-4-sulfonate, sodium 4-hydroxynaphthalene-1-sulfonate, 4-hydroxy-1-naphthalenesulfonic acid monosodium salt, sodium 4-hydroxy-1-naphthalenesulfonate, 1-naphthalenesulfonic acid, 4-hydroxy-, monosodium salt, 1-naphthalenesulfonic acid, 4-hydroxy-, sodium salt 1:1, 4-hydroxy-1-naphthalenesulfonic acid sodium salt, 84-87-7 parent
C1=CC=C2C(=C1)C(=CC=C2S(=O)(=O)[O-])O.[Na+]
246.212
246.21
Crystalline Powder

Chemical Identifiers

6099-57-6
246.212
IAAKNVCARVEIFS-UHFFFAOYSA-M
4626046
C1=CC=C2C(=C1)C(=CC=C2S(=O)(=O)[O-])O.[Na+]
C10H7NaO4S
MFCD00021518
1-naphthol-4-sulfonic acid sodium salt, sodium naphthol sulfonate, sodium 1-naphthol-4-sulfonate, sodium 4-hydroxynaphthalene-1-sulfonate, 4-hydroxy-1-naphthalenesulfonic acid monosodium salt, sodium 4-hydroxy-1-naphthalenesulfonate, 1-naphthalenesulfonic acid, 4-hydroxy-, monosodium salt, 1-naphthalenesulfonic acid, 4-hydroxy-, sodium salt 1:1, 4-hydroxy-1-naphthalenesulfonic acid sodium salt, 84-87-7 parent
sodium;4-hydroxynaphthalene-1-sulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0477

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions