missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-Naphthol-4-sulfonate 85.0+%, TCI America™

Catalog No. p-7133585
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N003325G 25 g
N0033100G 100 g
N0033500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N003325G Supplier TCI America Supplier No. N003325G
Only null left

Chemical Identifiers

CAS 6099-57-6
Molecular Formula C10H7NaO4S
Molecular Weight (g/mol) 246.212
MDL Number MFCD00021518
InChI Key IAAKNVCARVEIFS-UHFFFAOYSA-M
Synonym 1-naphthol-4-sulfonic acid sodium salt, sodium naphthol sulfonate, sodium 1-naphthol-4-sulfonate, sodium 4-hydroxynaphthalene-1-sulfonate, 4-hydroxy-1-naphthalenesulfonic acid monosodium salt, sodium 4-hydroxy-1-naphthalenesulfonate, 1-naphthalenesulfonic acid, 4-hydroxy-, monosodium salt, 1-naphthalenesulfonic acid, 4-hydroxy-, sodium salt 1:1, 4-hydroxy-1-naphthalenesulfonic acid sodium salt, 84-87-7 parent
PubChem CID 4626046
IUPAC Name sodium;4-hydroxynaphthalene-1-sulfonate
SMILES C1=CC=C2C(=C1)C(=CC=C2S(=O)(=O)[O-])O.[Na+]

Specifications

Quantity 25 g
Formula Weight 246.21
Percent Purity ≥85.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Naphthol-4-sulfonate
EINECSNumber (4)-0477
TSCA Yes