missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-Naphthol-3-sulfonate 80.0+%, TCI America™

Catalog No. N015025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N015025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N015025G Supplier TCI America Supplier No. N015025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13935-00-7
Molecular Formula C10H7NaO4S
Molecular Weight (g/mol) 246.212
MDL Number MFCD00064963
InChI Key IINOGGDSBSTVTO-UHFFFAOYSA-M
Synonym 4-Hydroxy-2-naphthalenesulfonic Acid Sodium Salt, Sodium 4-Hydroxy-2-naphthalenesulfonate, 1-Naphthol-3-sulfonic Acid Sodium Salt
PubChem CID 23663657
IUPAC Name sodium;4-hydroxynaphthalene-2-sulfonate
SMILES C1=CC=C2C(=C1)C=C(C=C2O)S(=O)(=O)[O-].[Na+]

Specifications

Quantity 25 g
Formula Weight 246.21
Percent Purity ≥80.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Naphthol-3-sulfonate
EINECSNumber (4)-0477
TSCA No