accessibility menu, dialog, popup

UserName

CAS RN 13935-00-7

IUPAC Name: sodium 4-hydroxynaphthalene-2-sulfonate
Synonyms: sodium 4-hydroxynaphthalene-2-sulfonate
Molecular Weight (g/mol): 246.21
Molecular Formula: C10H7NaO4S
InChi Key: IINOGGDSBSTVTO-UHFFFAOYSA-M
SMILES: [Na+].OC1=C2C=CC=CC2=CC(=C1)S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results