missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 1,2-Naphthoquinone-4-sulfonate 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 1 g |
| Formula Weight | 260.19 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Sodium 1,2-Naphthoquinone-4-sulfonate |
Sécurité et manipulation
| EINECSNumber | (4)-0569 |
| RTECSNumber | QK1490000 |
| TSCA | Yes |