missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 1.00 N Solution, APHA, Spectrum™ Chemical
Description
Spectrum™ Chemical Sodium Hydroxide, 1.00 N Solution, APHA is a standard solution of Caustic soda ideally suited for laboratory titrimetric techniques. You can rely on the quality of LabChem analytical test solutions for lot-to-lot reproducibility.
Specifications
Specifications
| CAS Max % | 0.0384, 0.9616 |
| Concentration | 1.00 N ± 0.03 N |
| Quantity | 200 L |
| Grade | Chemical Solution |
| Packaging | Plastic |
| Chemical Name or Material | Sodium Hydroxide Solution |