missing translation for 'onlineSavingsMsg'
Learn More

International Enviroguard™ SMS Lab Coats, Elastic Wrists, No Pockets

Breathable SMS lab coats feature three layers of protection against particulates, light liquid splashes, or sprays.

$117.01 - $129.33

Specifications

Product Type Controlled Environments Lab Coat
Gender Unisex
Material SMS
Color White
Closure Type Snaps
View More Specs

Products 6
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19472460
View Documents
International Enviroguard™
2246M
Medium
Case of 30 Each for $117.01
Only null left
 
19472461
View Documents
International Enviroguard™
2246L
Large
Case of 30 Each for $117.01
Only null left
 
19472462
View Documents
International Enviroguard™
2246XL
X-Large
Case of 30 Each for $117.01
Only null left
 
19472463
View Documents
International Enviroguard™
22462XL
2X-Large
Case of 30 Each for $120.09
Only null left
 
19472464
View Documents
International Enviroguard™
22463XL
3X-Large
Case of 30 Each for $123.17
Only null left
 
19472465
View Documents
International Enviroguard™
22464XL
4X-Large
Case of 30 Each for $129.33
Only null left
 
Description

Description

  • Disposable SMS lab coats are lightweight, breathable, and provide a particulate barrier
  • Durable, three-layer SMS fabric protects against light liquid splashes and sprays
  • Fabric keeps wearers cool for extended periods of time
  • No pockets, elastic wrists, folded collar
  • Recommended applications include pharmaceuticals, labs, healthcare, controlled environments, and more
Specifications

Specifications

Controlled Environments Lab Coat
SMS
Snaps
Single-use
Particulates
Serged
Unisex
White
Elastic
Non-sterile
None
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'