missing translation for 'onlineSavingsMsg'
Learn More

International Enviroguard™ SMS Lab Coats, Elastic Wrists, No Pockets

Catalog No. 19472462
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19472462 X-Large
19472460 Medium
19472461 Large
19472463 2X-Large
19472464 3X-Large
19472465 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19472462 Supplier International Enviroguard™ Supplier No. 2246XL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Breathable SMS lab coats feature three layers of protection against particulates, light liquid splashes, or sprays.

  • Disposable SMS lab coats are lightweight, breathable, and provide a particulate barrier
  • Durable, three-layer SMS fabric protects against light liquid splashes and sprays
  • Fabric keeps wearers cool for extended periods of time
  • No pockets, elastic wrists, folded collar
  • Recommended applications include pharmaceuticals, labs, healthcare, controlled environments, and more

Specifications

Product Type Controlled Environments Lab Coat
Size X-Large
Gender Unisex
Material SMS
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
Resistance Particulates
No. of Pockets None
Collar Type Folded
Seam Type Serged
For Use With (Application) Pharmaceutical, Healthcare, Controlled Environments
Show More Show Less