missing translation for 'onlineSavingsMsg'
Learn More

International Enviroguard™ SMS Lab Coats, Elastic Wrists, No Pockets

Catalog No. 19472460
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19472460 Medium
19472461 Large
19472462 X-Large
19472463 2X-Large
19472464 3X-Large
19472465 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19472460 Supplier International Enviroguard™ Supplier No. 2246M

Breathable SMS lab coats feature three layers of protection against particulates, light liquid splashes, or sprays.

  • Disposable SMS lab coats are lightweight, breathable, and provide a particulate barrier
  • Durable, three-layer SMS fabric protects against light liquid splashes and sprays
  • Fabric keeps wearers cool for extended periods of time
  • No pockets, elastic wrists, folded collar
  • Recommended applications include pharmaceuticals, labs, healthcare, controlled environments, and more

Specifications

Product Type Controlled Environments Lab Coat
Size Medium
Gender Unisex
Material SMS
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
Resistance Particulates
No. of Pockets None
Collar Type Folded
Seam Type Serged
For Use With (Application) Pharmaceutical, Healthcare, Controlled Environments
Show More Show Less