missing translation for 'onlineSavingsMsg'
Learn More

Guardian™ CP-25, Chemical Resistant Smooth Finish 25 mil Butyl Gloves

Heavyweight butyl gloves with fitted hand styling and a smooth finish offer protection against a wide range of hazardous chemicals.

$115.30 - $145.52

Specifications

Certifications/Compliance ASTM F739-05
Certifications/Compliance (Europe) CE
Color Black
Hand Specific
Latex Free Yes
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19321109
View Documents
Guardian™
52501
X-Small
Pair for $145.52
Only null left
 
19321110
View Documents
Guardian™
52502
Small
Pair for $145.52
Only null left
 
19321111
View Documents
Guardian™
52503
Medium
Pair for $115.30
Only null left
 
19321112
View Documents
Guardian™
52504
Large
Pair for $145.52
Only null left
 
19321113
View Documents
Guardian™
52505
X-Large
Pair for $115.30
Only null left
 
Description

Description

  • Butyl material resists oxidation, ozone corrosion and abrasions
  • Reusable and easy to clean
  • Fitted hand style with smooth texture
  • Protects against wide range of hazardous chemicals
  • 25 mil thickness, 14 in. length
  • Made in the USA
Specifications

Specifications

ASTM F739-05
Black
Yes
Powdered
0.63 mm
Gauntlet/Rolled
Unlined
14 in.
CE
Specific
Butyl
25 mil
Permeation resistance to most acids, aldehydes, alcohols, amides, bases, esters and ketones
Smooth
ISO 9001
35.5 cm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'