missing translation for 'onlineSavingsMsg'
Learn More

Guardian™ CP-25, Chemical Resistant Smooth Finish 25 mil Butyl Gloves

Catalog No. 19321109
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19321109 X-Small
19321110 Small
19321111 Medium
19321112 Large
19321113 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19321109 Supplier Guardian™ Supplier No. 52501

Heavyweight butyl gloves with fitted hand styling and a smooth finish offer protection against a wide range of hazardous chemicals.

  • Butyl material resists oxidation, ozone corrosion and abrasions
  • Reusable and easy to clean
  • Fitted hand style with smooth texture
  • Protects against wide range of hazardous chemicals
  • 25 mil thickness, 14 in. length
  • Made in the USA

Specifications

Certifications/Compliance ASTM F739-05
Certifications/Compliance (Europe) CE
Color Black
Hand Specific
Latex Free Yes
Material Butyl
Powdered Powdered
Size X-Small
Thickness (English) 25 mil
Thickness (Metric) 0.63 mm
For Use With (Application) Permeation resistance to most acids, aldehydes, alcohols, amides, bases, esters and ketones
Glove Cuff Style Gauntlet/Rolled
Grip Design Smooth
Lining Unlined
Manufacturing/QMS Audit Standards ISO 9001
Model 52501
Length (English) 14 in.
Length (Metric) 35.5 cm
Show More Show Less