missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guardian™ CP-25, Chemical Resistant Smooth Finish 25 mil Butyl Gloves
Heavyweight butyl gloves with fitted hand styling and a smooth finish offer protection against a wide range of hazardous chemicals.
Supplier: Guardian™ 52505
Description
- Butyl material resists oxidation, ozone corrosion and abrasions
- Reusable and easy to clean
- Fitted hand style with smooth texture
- Protects against wide range of hazardous chemicals
- 25 mil thickness, 14 in. length
- Made in the USA
Specifications
| ASTM F739-05 | |
| Black | |
| Yes | |
| Powdered | |
| 25 mil | |
| Permeation resistance to most acids, aldehydes, alcohols, amides, bases, esters and ketones | |
| Smooth | |
| ISO 9001 | |
| 14 in. |
| CE | |
| Specific | |
| Butyl | |
| X-Large | |
| 0.63 mm | |
| Gauntlet/Rolled | |
| Unlined | |
| 52505 | |
| 35.5 cm |