missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ SMER 3

Catalog No. 437510
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
437510 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 437510 Supplier R&D Systems™ Supplier No. 4375/10

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. Selective inhibitor of E3 ubiquitin ligase

Selective inhibitor of a yeast SCF family E3 ubiquitin ligase (SCFMet30) in vitro and in vivo. Induces the expression of MET genes; blocks cell proliferation. Enhancer of rapamycin (Cat. No. 1292).

Chemical Identifiers

CAS 67200-34-4
Molecular Formula C11H4N4O2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 224.18
Purity >98%
Molecular Weight (g/mol) 224.18
Chemical Name or Material SMER 3
Recommended Storage Store at +4°C