missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wanzl SmartCell™ Add on Unit with Baskets
SmartCell™ Shelving delivers industry-leading open wire storage that allows for easy identification of materials. Our quickly adjustable storage compartments increase the efficiency of both materials and staff management alike.
$1490.15 - $5096.09
Specifications
| Color | Chrome |
|---|---|
| Material | Steel |
| For Use With (Application) | Shelving |
| For Use With (Equipment) | SmartCell™ |
| Shelf Type | Add on Unit |
| Catalog Number | Mfr. No. | Length (English) | Includes | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Length (English) | Includes | Price | Quantity | ||||
|
11222003
|
Wanzl
THSC186A |
18 in. | 6 Baskets |
Each for $1,490.15
|
|
||||
|
11222005
|
Wanzl
THSC366A |
36 in. | 6 Baskets |
Each for $2,145.53
|
|
||||
|
11222007
|
Wanzl
THSC486A |
48 in. | 6 Baskets |
Each for $2,372.34
|
|
||||
|
11222015
|
Wanzl
THSC187A |
18 in. | 7 Baskets |
Each for $1,626.81
|
|
||||
|
11222017
|
Wanzl
THSC367A |
36 in. | 7 Baskets |
Each for $2,375.15
|
|
||||
|
11222019
|
Wanzl
THSC487A |
48 in. | 7 Baskets |
Each for $2,646.57
|
|
||||
|
11222009
|
Wanzl
THSC186A2 |
18 in. | 12 Baskets |
Each for $2,787.89
|
|
||||
|
11222011
|
Wanzl
THSC366A2 |
36 in. | 12 Baskets |
Each for $4,102.34
|
|
||||
|
11222013
|
Wanzl
THSC486A2 |
48 in. | 12 Baskets |
Each for $4,544.83
|
|
||||
|
11222021
|
Wanzl
THSC187A2 |
18 in. | 14 Baskets |
Each for $3,067.70
|
|
||||
|
11222023
|
Wanzl
THSC367A2 |
36 in. | 14 Baskets |
Each for $4,564.36
|
|
||||
|
11222025
|
Wanzl
THSC487A2 |
48 in. | 14 Baskets |
Each for $5,096.09
|
|
||||
Description
- Fine mesh wire baskets come in various sizes and are suitable for mounting on louvered panels or cantilever pedestal frames.
- Adjustable dividers provide endless variations to storage space and also create opportunities to implement lean inventory management practices, such as Kanban and PAR level systems.
- All baskets are plated in hospital-grade chrome and the framework is finished with industrial strength powder coat to enhance the durability and longevity of the system.
- Bottom dust shelf/cover provides a barrier and protection to bottom basket contents, maintaining Joint Commission standards.
- Wide variety of accessories available to customize the system to your application
Specifications
| Chrome | |
| Shelving | |
| Add on Unit |
| Steel | |
| SmartCell™ | |
| 74 in. |