missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wanzl SmartCell™ Add on Unit with Baskets
SmartCell™ Shelving delivers industry-leading open wire storage that allows for easy identification of materials. Our quickly adjustable storage compartments increase the efficiency of both materials and staff management alike.
Supplier: Wanzl THSC187A
Description
- Fine mesh wire baskets come in various sizes and are suitable for mounting on louvered panels or cantilever pedestal frames.
- Adjustable dividers provide endless variations to storage space and also create opportunities to implement lean inventory management practices, such as Kanban and PAR level systems.
- All baskets are plated in hospital-grade chrome and the framework is finished with industrial strength powder coat to enhance the durability and longevity of the system.
- Bottom dust shelf/cover provides a barrier and protection to bottom basket contents, maintaining Joint Commission standards.
- Wide variety of accessories available to customize the system to your application
Specifications
| Chrome | |
| Shelving | |
| 7 Baskets | |
| Add on Unit |
| Steel | |
| SmartCell™ | |
| 18 in. | |
| 74 in. |