Learn More
Thermo Scientific™ Smart -Vue Pro Sensors
Increase productivity in your lab with Thermo Scientific Smart Sensors.
$355.09 - $7,841.46
Specifications
Certifications/Compliance Smart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
For Use With (Application) Smart-Vue Pro Remote Monitoring System
For Use With (Equipment) Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
View More Specs
Products 23
Catalog Number Mfr. No. Type Calibration Range Cable Price Quantity & Availability  
Catalog Number Mfr. No. Type Calibration Range Cable Price Quantity & Availability  
SVSEN4CS001
View Documents Promotion Details
Thermo Scientific™
SVPSEN4CS001
Current Sensor Calibrated at 4, 8,12,16, 20 mA 4 to 20 mA, 0 to 5 V, or 0 to 10 V
Each for $1,327.35
Only null left
SVSEN4US001
View Documents Promotion Details
Thermo Scientific™
SVPSEN4US001
Current Sensor Not Calibrated 4 to 20 mA, 0 to 5 V, or 0 to 10 V
Each for $955.35
Only null left
SVSENRCS001
View Documents Promotion Details
Thermo Scientific™
SVPSENRCS001
Temp/RH Sensor Calibrated at +23°C / 50% RH
Each for $2,029.07
Only null left
SVSENRCS002
View Documents Promotion Details
Thermo Scientific™
SVPSENRCS002
Temp/RH Sensor Calibrated at +37°C / 90% RH
Each for $2,198.15
Only null left
SVSENRUS001
View Documents Promotion Details
Thermo Scientific™
SVPSENRUS001
Temp/RH Sensor Not Calibrated -40°C to +150°C, 0 to 99.9% RH
Each for $1,293.52
Only null left
SVSENTCE001
View Documents Promotion Details
Thermo Scientific™
SVPSENTCE001
Temperature Sensor Calibrated at -30°C, to +15°C, 0°C, 20°C, 40°C -40°C to +80°C 3.5 m
Each for $642.54
Only null left
SVSENTCS001
View Documents Promotion Details
Thermo Scientific™
SVPSENTCS001
Temperature Sensor Calibrated at -30°C, to +15°C, 0°C, 20°C, 40°C -40°C to +80°C 1 m
Each for $625.63
Only null left
SVSENTCS002
View Documents Promotion Details
Thermo Scientific™
SVPSENTCS002
Temperature Sensor Calibrated at 30°C, 60°C, 80°C,100°C, 105°C -40°C to +120°C 1 m
Each for $737.65
Only null left
SVSENTCS003
View Documents Promotion Details
Thermo Scientific™
SVPSENTCS003
PT100 Sensor Calibrated at -80°C, to +40°C, 0°C -100°C to +150°C 3.5 m
Each for $1,976.22
Only null left
SVSENTCS004
View Documents Promotion Details
Thermo Scientific™
SVPSENTCS004
PT100 Sensor Calibrated at -196°C, to +80°C, 0°C -200°C to +50°C 1.4 m
Each for $1,976.22
Only null left
SVSENTUE001
View Documents Promotion Details
Thermo Scientific™
SVPSENTUE001
Temperature Sensor Not Calibrated -40°C to +80°C 2.5 m
Each for $365.65
Only null left
SVSENTUS001
View Documents Promotion Details
Thermo Scientific™
SVPSENTUS001
PT100 Sensor Not Calibrated -40°C to +80°C 1 m
Each for $355.09
Only null left
SVSENTUS002
View Documents Promotion Details
Thermo Scientific™
SVPSENTUS002
Temperature Sensor Not Calibrated -40°C to +120°C, 0 to 99.9% RH 1 m
Each for $465.00
Only null left
SVSENTUS003
View Documents Promotion Details
Thermo Scientific™
SVPSENTUS003
PT100 Sensor Not Calibrated -100°C to +150°C 3.5 m
Each for $1,213.22
Only null left
SVSENTUS004
View Documents Promotion Details
Thermo Scientific™
SVPSENTUS004
PT100 Sensor Not Calibrated -200°C to +50°C 1.4 m
Each for $1,213.22
Only null left
SVSENCCS001
View Documents Promotion Details
Thermo Scientific™
SVPSENCCS001
CO2/Temp/RH Sensor 5% CO2, 37°C, 80% RH 0.5 to 9.9% CO2, 0°C to 50°C, 0 to 99% RH
Each for $7,841.46
Only null left
SVSENCUS001
View Documents Promotion Details
Thermo Scientific™
SVPSENCUS001
CO2/Temp/RH Sensor Not Calibrated 0.5 to 9.9% CO2, 0°C to 50°C, 0 to 99% RH
Each for $6,298.54
Only null left
SVSENDRY001
View Documents Promotion Details
Thermo Scientific™
SVPSENDRY001
Sensor Cable Not Calibrated N/A 2.9 m
Each for $372.00
Only null left
SVSENPCS001
View Documents Promotion Details
Thermo Scientific™
SVPSENPCS001
Differential Pressure Sensor 0, 15, 25, 50 Pa -500 Pa to +500 Pa. (-2 to +2 in./-5 to +5 cm in H2O)
Each for $3,212.67
Only null left
SVSENPUS001
View Documents Promotion Details
Thermo Scientific™
SVPSENPUS001
Differential Pressure Sensor Not Calibrated -500 Pa to +500 Pa. (-2 to +2 in./-5 to +5 cm in H2O)
Each for $2,196.15
Only null left
SVWENTCS001
View Documents Promotion Details
Thermo Scientific™
SVPWSENTCS001
Wireless Temp Sensor 5°C -40°C to +85°C N/A
Each for $910.96
Only null left
SVWENTCS002
View Documents Promotion Details
Thermo Scientific™
SVPWSENTCS002
Wireless Temp Sensor -80°C -200°C to +50°C N/A
Each for $1,335.80
Only null left
SVWENTCS003
View Documents Promotion Details
Thermo Scientific™
SVPWSENTCS003
Wireless Temp Sensor 5°C -50°C to +200°C N/A
Each for $1,796.57
Only null left
Description
Description
Increase productivity and ensure the integrity of your samples and research, align with 21 CFR Part 11, and continuously monitor and track conditions using Smart-Vue Pro sensors. Monitor temperatures from -200°C to +150°C, current, and relative humidity. With off the shelf calibrated sensors you can be confident in the integrity of your laboratory equipment.
Multiple parameters
  • Temperature (-200 /+150°C)
  • Temperature and relative humidity
  • 0-5 V / 0-10 V / 4-20mA
  • Dry contact
SMART Calibration (for SMART Sensors Only)
  • Off the shelf Calibrated sensors 
  • Calibration coefficients stored in sensor memory to simplify recalibration process
  • No data loss or down-time for calibration
  • Data and certificates accessible easily
  • Calibration in-house laboratory: ISO 17025 accredited (COFRAC) and NIST-traceable
  • (Optional)On Site Calibration
Modular sensor configuration
  • SMART Sensors enable Plug and play feature 
  • Module Sensor auto-detection
Specifications
Specifications
Smart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Smart-Vue Pro Remote Monitoring System
Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module