missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SLU-PP-332
Description
SLU-PP-332 is a potent and selective pan estrogen related receptor (ERR) agonist (EC50 values are 98 nM, 230 nM and 430 nM for ERRα, ERRβ and ERRγ respectively). In a mouse model of metabolic syndrome SLU-PP-332 increases energy expenditure and fatty acid oxidation, decreases fat mass, and improves insulin sensitivity. Also increases mitochondrial respiration in vitro.
Specifications
Specifications
| Chemical Name or Material | (E)-4-Hydroxy-N'-(naphthalen-2-ylmethylene)benzohydrazide |
| CAS | 303760-60-3 |
| Quantity | 50 mg |
| Target | Estrogen and Related Receptor Agonists |
| Molecular Formula | C18H14N2O2 |
| Purity | 0.98 |