Learn More
Thermo Scientific™ Racks for Thermo Scientific™ 5 Shelf TSX/TDE Series Freezers
Maximize tube storage capacity per freezer with a racking solution for five inner-door, ultra-low temperature freezers.
$529.57 - $925.26
Products 19
Catalog Number Mfr. No. Holds Positions For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. Holds Positions For Use With (Equipment) Price Quantity & Availability  
1950637
View Documents Promotion Details
Thermo Scientific™
1950637
Microplates 16 Plates/Rack EXF240, HFU240B (5 shelf) (13 cu.ft.)
Each for $714.83
Only null left
1950638
View Documents Promotion Details
Thermo Scientific™
1950638
Microplates 24 Plates/Rack EXF320, EXF400, HFU320B, HFU400B (5 shelf) (17.3, 23 cu.ft.)
Each for $628.87
Only null left
1950639
View Documents Promotion Details
Thermo Scientific™
1950639
Microplates 28 Plates/Rack Revco UxF, HERAFreeze HFU-T (14.9, 19.4, 24.1, 28.8, 33.5 cu.ft.)
Each for $682.64
Only null left
1950792
View Documents Promotion Details
Thermo Scientific™
1950792
Tubes 96/Box EXF240, HFU240B (13 cu.ft.)
Each for $562.66
Only null left
1950794
View Documents Promotion Details
Thermo Scientific™
1950794
Matrix 2D Tubes (1 mL) 18 Boxes/Rack EXF320, EXF400, HFU320B, HFU400B (17.3, 23 cu.ft.)
Each for $622.66
Only null left
1950798
View Documents Promotion Details
Thermo Scientific™
1950798
Matrix 2D Tubes (1 mL) 21 Boxes/Rack Revco UxF, HERAFreeze HFU-T (14.9, 19.4, 24.1, 28.8, 33.5 cu.ft.)
Each for $925.26
Only null left
398321
View Documents Promotion Details
Thermo Scientific™
398321
Microplates with Lids 70 Plates/Rack Upright Freezers (13 cu.ft.)
Each for $628.87
Only null left
398323
View Documents Promotion Details
Thermo Scientific™
398323
Microplates with Lids 84 Plates/Rack Upright Freezers (17.3, 23 cu.ft.)
Each for $724.45
Only null left
13994158
View Documents Promotion Details
Thermo Scientific™
398324
Boxes (2 in.) 12 Boxes/Rack Upright Freezers (13 cu.ft.)
Each for $546.13
Only null left
398325
View Documents Promotion Details
Thermo Scientific™
398325
Boxes (3 in.) 9 Boxes/Rack Upright Freezers (13 cu.ft.)
Each for $589.26
Only null left
398326
View Documents Promotion Details
Thermo Scientific™
398326
Boxes (2 in.) 16 Boxes/Rack Upright Freezers (17.3, 23 cu.ft.)
Each for $628.87
Only null left
398330
View Documents Promotion Details
Thermo Scientific™
398330
Boxes (3 in.) 12 Boxes/Rack Upright Freezers (17.3, 23 cu.ft.)
Each for $599.89
Only null left
398331
View Documents Promotion Details
Thermo Scientific™
398331
Boxes (2 in.) 16 Boxes/Rack Upright Freezers (17.3, 23 cu.ft.)
Each for $599.89
Only null left
398332
View Documents Promotion Details
Thermo Scientific™
398332
Boxes (2 in.) 12 Boxes/Rack Upright Freezers (13 cu.ft.)
Each for $562.66
Only null left
398333
View Documents Promotion Details
Thermo Scientific™
398333
Microplates 66 Plates/Rack Upright Freezers (17.3, 23, 28.8 cu.ft.)
Each for $693.00
Only null left
398334
View Documents Promotion Details
Thermo Scientific™
398334
Microplates 44 Plates/Rack Upright Freezers (13 cu.ft.)
Each for $564.74
Only null left
15467446
View Documents Promotion Details
Thermo Scientific™
920099
Boxes (3 in.) 9 Boxes/Rack Upright Freezers (13 cu.ft.)
Each for $529.57
Only null left
398335
View Documents Promotion Details
Thermo Scientific™
398335
Microplates 30 Plates/Rack Upright Freezers (17.3, 23, 28.8 cu.ft.)
Each for $628.87
Only null left
398327
View Documents Promotion Details
Thermo Scientific™
398327
Boxes (2 in.) 12 Boxes/Rack Upright Freezers (17.3, 23 cu.ft.)
Each for $628.87
Only null left
Description
Description
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for TSX/TDE 5 shelf Series Freezers as well as well as previous generation Revco™ ExF, DxF and HERAfeeze™ HFU B Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
Thermo Scientific™ freezer racking and tubing solutions offer a convenient, streamlined process to maximize sample storage in your laboratory. Our racking solutions are designed to hold tubes ranging from 200μL up to 5.0mL, offering maximized sample storage capacity.
  • With sliding drawer racks
  • Adjustable side access box racks
  • Standard/deepwell microplate racks
  • Warranty varies by country, please contact us for details