Learn More
United Scientific™ Slide Mailer, Cardboard
Ideal for Holding and transferring microscope slides.
Supplier: United Scientific™ 62600
Description
- Designed as a convenient solution for holding and transferring microscope slides.
- Fold-over cover helps protect the slides.
- Accommodates microscope slides in two sizes:
- 75.0 x 25.0 x 0.8 to 1.2 mm
- 76.0 x 26.0 x 0.8 to 1.2 mm
- Heavy duty cardboard
- Thumb cuts for easy removal of slides
- Flat writing surface on inside cover for specimen identification
Specifications
| Slide Mailer | |
| 1 | |
| Cardboard |