Learn More
United Scientific™ Slide Mailer, Cardboard
Catalog No. p-200025638
Change view
Click to view available options
Slide Capacity:
1
2
4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Slide Capacity
S012580 1
S012579 2
S012578 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S012580 Supplier United Scientific™ Supplier No. 62600
Only null left
Ideal for Holding and transferring microscope slides.
  • Designed as a convenient solution for holding and transferring microscope slides.
  • Fold-over cover helps protect the slides.
  • Accommodates microscope slides in two sizes:
    • 75.0 x 25.0 x 0.8 to 1.2 mm
    • 76.0 x 26.0 x 0.8 to 1.2 mm
  • Heavy duty cardboard
  • Thumb cuts for easy removal of slides
  • Flat writing surface on inside cover for specimen identification
Specifications
Type Slide Mailer
Slide Capacity 1
Material Cardboard