Learn More
United Scientific™ Slide Mailer, Cardboard
Ideal for Holding and transferring microscope slides.
Supplier:  United Scientific™ 62601
 View more versions of this product
Catalog No. S012579
Only null left
Explore available promotions
Description
Description
  • Designed as a convenient solution for holding and transferring microscope slides.
  • Fold-over cover helps protect the slides.
  • Accommodates microscope slides in two sizes:
    • 75.0 x 25.0 x 0.8 to 1.2 mm
    • 76.0 x 26.0 x 0.8 to 1.2 mm
  • Heavy duty cardboard
  • Thumb cuts for easy removal of slides
  • Flat writing surface on inside cover for specimen identification
Specifications
Specifications
Slide Mailer
2
Cardboard
Product Suggestions
Product Suggestions