missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals SKF-96365 hydrochloride, 99+%

Inhibits receptor-mediated calcium influx

Chemical Identifiers

CAS 130495-35-1
Molecular Formula C22H26N2O3·HCl, C22H27ClN2O3
Synonym 1-[2-(4-Methoxyphenyl)-2-[3-(4-methoxyphenyl)propoxy]ethyl]imidazole
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ60937MA
View Documents Promotion Details
Thermo Scientific Chemicals
J60937MA
10 mg N/A
Only null left
 
Description

Description

Inhibits receptor-mediated calcium influx

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

130495-35-1
C22H26N2O3·HCl, C22H27ClN2O3
1-[2-(4-Methoxyphenyl)-2-[3-(4-methoxyphenyl)propoxy]ethyl]imidazole

Specifications

White
130495-35-1
402.92g/mol
Soluble to 100mM in water
Powder
[H+].[Cl-].COC1=CC=C(CCCOC(CN2C=CN=C2)C2=CC=C(OC)C=C2)C=C1
402.92
Inhibits receptor-mediated calcium influx
C22H26N2O3·HCl, C22H27ClN2O3
1-[2-(4-Methoxyphenyl)-2-[3-(4-methoxyphenyl)propoxy]ethyl]imidazole
Signal Transduction Reagent
FWLPKVQUECFKSW-UHFFFAOYNA-N
hydrogen 1-[2-(4-methoxyphenyl)-2-[3-(4-methoxyphenyl)propoxy]ethyl]-1H-imidazole chloride
≥99%
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only