missing translation for 'onlineSavingsMsg'
Learn More

Simulated Intestinal Fluid TS, Ricca Chemical

$307.00 - $307.00

Specifications

Chemical Name or Material Simulated Intestinal Fluid
Color Undesignated
Name Note Test Solution
CAS 7732-18-5,8049-47-6,7778-77-0,1310-73-2
CAS Min % 96.0,0.84,0.57,0.1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
7109-16
View Documents
Ricca Chemical Company
RI710916
500 mL
Each for $307.00
Only null left
 
Description

Description

Specifications

Specifications

Simulated Intestinal Fluid
Test Solution
96.0,0.84,0.57,0.1
Laboratory
6.7 to 6.9
Undesignated
7732-18-5,8049-47-6,7778-77-0,1310-73-2
100.0,0.87,0.59,0.11
Natural Poly Bottle
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'