missing translation for 'onlineSavingsMsg'
Learn More

Simulated Intestinal Fluid TS, Ricca Chemical

Catalog No. 710916
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
710916 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 710916 Supplier Ricca Chemical Company Supplier No. RI710916
Only null left

Specifications

Chemical Name or Material Simulated Intestinal Fluid
Color Undesignated
Name Note Test Solution
CAS 7732-18-5, 8049-47-6, 7778-77-0, 1310-73-2
CAS Min % 96.0, 0.84, 0.57, 0.1
CAS Max % 100.0, 0.87, 0.59, 0.11
Grade Laboratory
Packaging Natural Poly Bottle
pH 6.7 to 6.9
Quantity 500 mL
Physical Form Liquid
Show More Show Less