missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific T100 BIOTUBE™ Tubes

T100 BIOTUBE™ Tubes are ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies.

$366.21 - $376.29

Specifications

Autoclavable Autoclavable
Capacity (Metric) 1.2 mL
Material Polypropylene
For Use With (Application) Ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting, pharmaceutical quality control, receptor binding assays, RIA and EIA
For Use With (Equipment) T100 Biotube
View More Specs

Products 2
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
05-500-18
View Documents
Simport™ Scientific
T10025
Strips of 8 Tubes
Case of 600 Each for $366.21
Only null left
 
05-500-16
View Documents
Simport™ Scientific
T10020
Individual Tubes without Writing Surface
Case of 4800 Each for $376.29
Only null left
 
Description

Description

T100 BIOTUBE™ Tubes are ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies.

  • Polypropylene construction
  • Caps sold separately
  • Gross volume: 1.2 mL, with cap : 1.1 mL
  • Used for procedures requiring low surface tension such as protein, nucleic acid work
  • Designed to avoid potentially harmful lubricants while minimizing liquid retention
  • Applications: Ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting, pharmaceutical quality control, receptor binding assays, RIA and EIA
Specifications

Specifications

Autoclavable
Polypropylene
T100 Biotube
Non-sterile
1.2 mL
Ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting, pharmaceutical quality control, receptor binding assays, RIA and EIA
600/Cs.
2000 x g
Videos
SDS
Documents

Documents