missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Simport™ Scientific T100 BIOTUBE™ Tubes
Description
T100 BIOTUBE™ Tubes are ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies.
- Polypropylene construction
- Caps sold separately
- Gross volume: 1.2 mL, with cap : 1.1 mL
- Used for procedures requiring low surface tension such as protein, nucleic acid work
- Designed to avoid potentially harmful lubricants while minimizing liquid retention
- Applications: Ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting, pharmaceutical quality control, receptor binding assays, RIA and EIA
Specifications
Specifications
| Autoclavable | Autoclavable |
| Capacity (Metric) | 1.2 mL |
| Material | Polypropylene |
| For Use With (Application) | Ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting, pharmaceutical quality control, receptor binding assays, RIA and EIA |
| For Use With (Equipment) | T100 Biotube |
| Includes | Strips of 8 Tubes |
| No. per Pack | 600/Cs. |
| Sterility | Non-sterile |
| Max. RCF | 2000 x g |