missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific CytoSep™ Double Sample Chambers

Catalog No. 22045325
Change view
Click to view available options
Type:
Double Sample Chamber
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22045325 Double Sample Chamber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22045325 Supplier Simport™ Scientific Supplier No. M967 20FW
Only null left

CytoSep™ Double Sample Chambers for use with Wescor Cyto-System.

  • Available in white and tan

Specifications

Color White
For Use With (Equipment) Wescor Cyto-System
Includes White Filter Paper and Cap
Type Double Sample Chamber