Learn More
Simport™ Scientific CytoSep™ Funnels for Shandon™ Cytospin 4™ Centrifuges
CytoSep™ Funnels for Shandon™ Cytospin 4™ Centrifuges for use with Thermo Scientific™ Shandon Cytospin 4 Centrifuges.
Supplier:  Simport™ Scientific M9651
 View more versions of this product
Catalog No. 22-045-312
Only null left
Explore available promotions
Description
Description
  • All plastic disposable sample chambers do not need metal clips
  • Available with or without pre-attached filter card
Specifications
Specifications
Shandon Cytospin 4
Single Funnel Only, All Plastic
Product Suggestions
Product Suggestions