Learn More
Simport™ Scientific CytoSep™ Funnels for Shandon™ Cytospin 4™ Centrifuges
CytoSep™ Funnels for Shandon™ Cytospin 4™ Centrifuges for use with Thermo Scientific™ Shandon Cytospin 4 Centrifuges.
Supplier: Simport™ Scientific M9641D
Description
- All plastic disposable sample chambers do not need metal clips
- Available with or without pre-attached filter card
Specifications
| Shandon Cytospin 4 | |
| Double Cytology Funnel Only |