Learn More
Simport™ Scientific CytoSep™ Funnels for Shandon™ Cytospin 4™ Centrifuges
Description
- All plastic disposable sample chambers do not need metal clips
- Available with or without pre-attached filter card
Specifications
Specifications
| Type | Double Funnel w/ White Filter and Cap |
| For Use With (Equipment) | Shandon Cytospin 4 |