missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific Polyethylene Flange Plug Caps

These Flange Plug Caps have two flexible flanges to ensure a leakproof seal. They will fit into test tubes and also in round cuvettes and centrifuge tubes.

Supplier:  Simport™ Scientific T4043R

 View more versions of this product

Catalog No. 22045564


Only null left

Description

Description

  • Made of polyethylene
  • Easy-on, easy-off design on 12 m and 16 mm OD tubes
  • Protects from aerosols of highly infectious microorganisms
  • Available in 6 colors
Specifications

Chemical Identifiers

Polyethylene

Specifications

Flange Plug Cap
Polyethylene
0.47 in.
12 mm Tubes
Test tubes, round cuvettes, centrifuge tubes, two flexible flanges
Red
12 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions