missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Simazine, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC S3280
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| Simazine | 122-34-9 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Simazine | |
| 122-34-9,67-64-1 | |
| Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity. | |
| For US EPA Method 619 | |
| GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
| ODCWYMIRDDJXKW-UHFFFAOYSA-N | |
| Acetone | |
| 201.658 | |
| 5216 |
| 2mL amber prescored glass ampule | |
| 0.1,99.9 | |
| 0.1,99.9 | |
| C7H12ClN5 | |
| Certified Reference Material | |
| CCNC1=NC(=NC(=N1)Cl)NCC | |
| 6-chloro-2-N,4-N-diethyl-1,3,5-triazine-2,4-diamine | |
| 1 mL | |
| CHEBI:27496 |
Chemical Identifiers
| 122-34-9 | |
| 201.658 | |
| 5216 | |
| 6-chloro-2-N,4-N-diethyl-1,3,5-triazine-2,4-diamine |
| C7H12ClN5 | |
| ODCWYMIRDDJXKW-UHFFFAOYSA-N | |
| CHEBI:27496 | |
| CCNC1=NC(=NC(=N1)Cl)NCC |
Safety and Handling
Highly flammable liquid and vapor. Toxic if inhaled. Causes damage to organs.
DOTInformation : Nonhazardous
Recommended Storage : Freezer
Product intended for laboratory use only