missing translation for 'onlineSavingsMsg'
Learn More

Silver Standard, 1mL = 1mg Ag, 1000ppm Ag, Ricca Chemical

$1120.00 - $1120.00

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
71004
View Documents
Thermo Fisher Scientific
71004
120 mL Amber Poly
Each for $1,120.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7761-88-8
169.87
SQGYOTSLMSWVJD-UHFFFAOYSA-N
CHEBI:32130
[Ag+].[O-][N+]([O-])=O
AgNO3
MFCD00003414
24470
silver(1+) nitrate

Specifications

Amber Poly
1000ppm Ag
Liquid
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver(1+) nitrate
120 mL
CHEBI:32130
7761-88-8,7697-37-2,7732-18-5
1 mL = 1 mg Ag
AgNO3
Colorless
[Ag+].[O-][N+]([O-])=O
169.87
24470
Silver Standard
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'