missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, 0.153 N, Ricca Chemical

Catalog No. 697532
Change view
Click to view available options
Quantity:
1 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
697532 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 697532 Supplier Ricca Chemical Company Supplier No. 697532

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5 , 7761-88-8
Molecular Weight (g/mol) 169.87 g/mol

Specifications

Color Colorless
Physical Form Liquid
Quantity 1 L
Grade Laboratory
Packaging Amber Glass Bottle
Chemical Name or Material Silver nitrate solution, 0.153N