missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, Certified, 10.0% (w/v) ±0.3%, LabChem™

Catalog No. p-5388214
Change view
Click to view available options
Quantity:
125 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC225507 125 mL
LC225501 500 mL
LC225502 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC225507 Supplier LabChem Supplier No. LC225507
Only null left

Chemical Identifiers

CAS 7761-88-8 , 7732-18-5
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Color Colorless
CAS Max % 90, 10
Physical Form Liquid
Quantity 125 mL
Linear Formula AgNO3
Solubility Information Soluble in water
Formula Weight 169.87
Percent Purity 10.0% ±0.3% (w/v)
Grade Certified
Identification Passes Test
Packaging Amber Glass
Specific Gravity 1.4g/mL
Decomposition Information Nitrogen oxides; Thermal decomposition generates: Corrosive vapors
Density 1.4g/mL
Chemical Name or Material Silver Nitrate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Do not breathe mist, spray, vapors.
Avoid release to the environment.
Wear eye protection, protective clothing, protective gloves, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature