missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, Certified, 0.0250N ±0.0003N (0.025M), LabChem™
Specifications
Specifications
| Boiling Point | 100°C |
| Color | Colorless |
| CAS Max % | 99.58, 0.42 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Linear Formula | AgNO3 |
| Solubility Information | Soluble in water |
| Formula Weight | 169.87 |
| Grade | Certified |
| Concentration or Composition (by Analyte or Components) | 0.0250N ±0.0003N (0.025M) |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Solution is not hazardous. |
| Precautionary Statement | GHS P Statement If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes. If swallowed, get medical attention. |
| Recommended Storage | Room Temperature |