missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, ACS Grade, ≥ 99.0%, LabChem™
Supplier: LabChem LC225001
Specifications
| 444°C | |
| 212°C | |
| White | |
| Solid | |
| AgNO3 | |
| MFCD00003414 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| silver(1+) nitrate | |
| 24470 | |
| 169.87 | |
| ACS | |
| Amber Glass | |
| Nitrogen oxides |
| Silver Nitrate | |
| 7761-88-8 | |
| 100 | |
| 500 g | |
| AgNO3 | |
| UN1493 | |
| Soluble in water | |
| [Ag+].[O-][N+]([O-])=O | |
| 169.87 | |
| CHEBI:32130 | |
| ≥99.0% | |
| Passes Test | |
| 4352kg/m3 | |
| 4352kg/m3 |
Chemical Identifiers
| 7761-88-8 | |
| 169.87 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| 24470 | |
| silver(1+) nitrate |
| AgNO3 | |
| MFCD00003414 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| CHEBI:32130 | |
| [Ag+].[O-][N+]([O-])=O |
Safety and Handling
GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat, open flames, sparks.
No smoking.
Keep/Store away from combustible materials.
Take any precaution to avoid mixing with combustibles.
Do not breathe dust.
Do not eat, drink or smoke when usi
Danger
EINECSNumber : 231-853-9
Recommended Storage : Room Temperature