missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, ACS Grade, ≥ 99.0%, LabChem™

Catalog No. p-5379498
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC225009 100 g
LC225001 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC225009 Supplier LabChem Supplier No. LC225009
Only null left

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Boiling Point 444°C
Melting Point 212°C
Color White
CAS Max % 100
Physical Form Crystals
Quantity 100 g
Linear Formula AgNO3
UN Number UN1493
Solubility Information Soluble in water
Formula Weight 169.87
Percent Purity ≥99.0%
Grade ACS
Identification Passes Test
Packaging Amber Glass
Specific Gravity 4352kg/m3
Decomposition Information Nitrogen oxides
Density 4,352kg/m3
Chemical Name or Material Silver Nitrate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat, open flames, sparks.
No smoking.
Keep/Store away from combustible materials.
Take any precaution to avoid mixing with combustibles.
Do not breathe dust.
Do not eat, drink or smoke when using this product.
Wear eye protection, face protection, protective gloves, protective clothing.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 231-853-9
Recommended Storage Room Temperature