missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate, Acculute Standard Volumetric Solution, Final Concentration 0.1N

CAS: 7761-88-8 | AgNO3 | 169.87 g/mol

$228.07 - $228.07

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA88614LQ
View Documents
Thermo Scientific Chemicals
088614LQ
1 Unit
Each for $228.07
Only null left
 
AA88614LS
View Documents
Thermo Scientific Chemicals
088614LS
6 Units N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Silver nitrate 0.1N standard volumetric solution is used for volumetric analysis in laboratories. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7761-88-8
169.87
SQGYOTSLMSWVJD-UHFFFAOYSA-N
CHEBI:32130
[Ag+].[O-][N+]([O-])=O
AgNO3
MFCD00003414
24470
silver(1+) nitrate

Specifications

7761-88-8
Liquid
MFCD00003414
Miscible with water, alcohol and aqueous ammonia
[Ag+].[O-][N+]([O-])=O
169.87
24470
169.87
Silver nitrate
23 hPa (17mm Hg) at 20°C
AgNO3
Colorless
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver(1+) nitrate
1 Unit
CHEBI:32130
Silver nitrate: 40%; Water: 60%
Videos
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H271-H314-H335

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 231-853-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only