missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, (1mL = 5mg Cl-), Certified, 0.141N ±0.002N (0.141M), LabChem™
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 97.6, 2.4 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Linear Formula | AgNO3 |
| Solubility Information | Soluble in water |
| Formula Weight | 169.87 |
| Grade | Certified |
| Concentration or Composition (by Analyte or Components) | 0.141N ±0.002N (0.141M) |
| Identification | Passes Test |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Causes skin irritation and serious eye irritation. Harmful to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Wear eye protection, protective gloves. Wash exposed skin thoroughly after handling. Avoid release to the environment. If on skin: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. If skin irritation occurs: Get medical advice/attention. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |