missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, (1mL = 10mg Cl-), Certified, 0.282N ±0.003N (0.282M), LabChem™

Catalog No. LC227251
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC227251 500 mL
LC227252 1 L
LC227254 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC227251 Supplier LabChem Supplier No. LC227251
Only null left

Chemical Identifiers

CAS 7761-88-8 , 7732-18-5
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Color Colorless
CAS Max % 95.21, 4.79
Physical Form Liquid
Quantity 500 mL
Linear Formula AgNO3
Solubility Information Soluble in water
Formula Weight 169.87
Grade Certified
Concentration or Composition (by Analyte or Components) 0.282N ±0.003N (0.282M)
Identification Passes Test
Packaging Amber Glass
Specific Gravity 1g/mL
Decomposition Information Nitrogen oxides
Density 1g/mL
Chemical Name or Material Silver Nitrate, (1mL = 10mg Cl-)
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Harmful to aquatic life.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature