missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, 10% (w/v) (100 g/L), Ricca Chemical
For Chemical Testing
$319.57 - $1749.97
Chemical Identifiers
| Molecular Formula | AgNO3 |
|---|---|
| Molecular Weight (g/mol) | 169.87 |
| MDL Number | MFCD00003414 |
| InChI Key | SQGYOTSLMSWVJD-UHFFFAOYSA-N |
| Synonym | silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol |
| PubChem CID | 24470 |
| ChEBI | CHEBI:32130 |
| IUPAC Name | silver(1+) nitrate |
| SMILES | [Ag+].[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
6830-4
|
Ricca Chemical Company
68304 |
120 mL | Amber Poly Bottle |
Each for $319.57
|
|
||||
|
6830-16
|
Ricca Chemical Company
683016 |
500 mL | Amber Poly Bottle |
Each for $1,017.93
|
|
||||
|
683032
|
Ricca Chemical Company
683032 |
1 L | Amber Poly Bottle |
Each for $1,749.97
|
|
||||
Chemical Identifiers
| AgNO3 | |
| MFCD00003414 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| CHEBI:32130 | |
| [Ag+].[O-][N+]([O-])=O |
| 169.87 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| 24470 | |
| silver(1+) nitrate |
Specifications
| 7732-18-5 | |
| MFCD00003414 | |
| [Ag+].[O-][N+]([O-])=O | |
| 169.87 | |
| Laboratory | |
| ASTM for Sulfate | |
| 120 mL |
| AgNO3 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| silver(1+) nitrate | |
| 10% | |
| Amber Poly Bottle | |
| ∼6 | |
| Silver Nitrate |