missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, 0.171 N Standardized Solution, Spectrum™ Chemical

Catalog No. p-7166399
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Glass Bottle
Catalog No. Quantity Packaging
18613177 500 mL Amber Glass Bottle
18613179 4 L Glass Bottle
18613180 20 L Cubitainer
3 options

Catalog No. 18613177

Supplier: Spectrum Chemical Mfg Cor S166500ML

Only null left

Chemical Identifiers

CAS 7761-88-8 , 7732-18-5
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Quantity 500 mL
Concentration or Composition (by Analyte or Components) 0.171 N ± 0.005 N
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.