missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, 0.100 Normal USP Testing Method, Reagents

Catalog No. 01336374
Change view
Click to view available options
Quantity:
1 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336374 1 L
01336373 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336374 Supplier Labchem Inc Supplier No. C73031001C
Only null left

Silver Nitrate, 0.100 Normal USP Testing Method, Meets USP specifications, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7761-88-8
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Nitric Acid silver salt
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
Physical Form Liquid
Quantity 1 L
Grade USP
Packaging Glass Bottle
Specific Gravity 1.0145
Chemical Name or Material Silver Nitrate, 0.100 Normal USP Testing Method