missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, 0.1 N, Standardized Solution, Spectrum™ Chemical

Catalog No. 18613181
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613181 500 mL Amber Glass Bottle
18613182 1 L Amber Glass Bottle
18613183 4 L Poly Bottle
18613184 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18613181 Supplier Spectrum Chemical Mfg Cor Supplier No. S163500MLGL
Only null left

Chemical Identifiers

CAS 7761-88-8 , 7732-18-5
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Quantity 500 mL
Concentration or Composition (by Analyte or Components) 0.100 N ± 0.0005 N
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.