missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, 0.0500 Normal (N/20), Reagents

Catalog No. 01333628
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Cubitainer
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01333628 20 L Cubitainer
01333629 1 L Glass Bottle
01333627 4 L Glass Bottle
01333626 4 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01333628 Supplier Labchem Inc Supplier No. CS14510020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Silver Nitrate, 0.0500 Normal (N/20), Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7761-88-8
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Nitric Acid silver salt
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
Physical Form Liquid
Quantity 20 L
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.0075
Chemical Name or Material Silver Nitrate, 0.0500 Normal (N/20)