missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, 0.0342 Normal, Reagents

Catalog No. 01333669
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333669 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01333669 Supplier Labchem Inc Supplier No. CS13990020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Silver Nitrate, 0.0342 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7761-88-8
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Nitric Acid silver salt
PubChem CID 24470
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
Physical Form Liquid
Quantity 20 L
Linear Formula AgNO3
Solubility Information Miscible in Water
Formula Weight 169.87 g/mol
Grade Reagent
Odor Odorless
Packaging Cubitainer
Appearance Colorless Liquid
Specific Gravity 1.0052
Chemical Name or Material Silver Nitrate, 0.0342 Normal
Show More Show Less
Recommended Storage Room Temperature