missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, 0.001 Normal, Volumetric, Reagents

Catalog No. 01333935
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333935 1 L
01333934 4 L
01333933 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01333935 Supplier Labchem Inc Supplier No. CS0150001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Silver Nitrate, 0.001 Normal, Volumetric, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7761-88-8
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Nitric Acid silver salt
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
Physical Form Liquid
Quantity 1 L
Solubility Information Miscible in Water
PubChem CID 24470
Formula Weight 169.87 g/mol
Grade Reagent
Packaging Glass Bottle
Appearance Colorless Liquid
Specific Gravity 1.0001
Chemical Name or Material Silver Nitrate, 0.001 Normal, Volumetric
Show More Show Less
Recommended Storage Room Temperature